Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198625.1 | NP_941027.1 | 13 - 14 | 1599 | 59 |
GenBank mRNA | AK052172.1 | - | 3 - 4 | 420 | 59 |
AK137832.1 | - | 13 - 14 | 1583 | 59 | |
BC060632.1 | - | 13 - 14 | 1599 | 59 |