Assay Design
Gene Transcripts
- Gene Symbol
- Coq9
- Entrez Gene ID
-
67914
- Gene Name
- coenzyme Q9 homolog (yeast)
- Gene Aliases
-
2310005O14Rik, C78387
- Chromosome Location
-
Chr.8: 94838417 - 94854895 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK009179.1 |
- |
1 - 2 |
92 |
146 |
AK154343.1 |
- |
1 - 2 |
104 |
148 |
AK158979.1 |
- |
1 - 2 |
98 |
148 |
View Full Product Details