Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170691.1 | NP_001164162.1 | 4 - 5 | 409 | 89 |
NM_178736.5 | NP_848851.3 | 4 - 5 | 414 | 89 | |
GenBank mRNA | AK029044.1 | - | 4 - 5 | 385 | 89 |
AK036607.1 | - | 4 - 5 | 392 | 89 | |
AK036746.1 | - | 4 - 5 | 354 | 89 | |
AK041137.1 | - | 4 - 5 | 315 | 89 | |
AK082730.1 | - | 3 - 4 | 181 | 89 | |
AK084950.1 | - | 4 - 5 | 414 | 89 | |
BC079654.1 | - | 4 - 5 | 375 | 89 |