Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001102615.1 | NP_001096085.1 | 17 - 18 | 2430 | 83 |
GenBank mRNA | AK155544.1 | - | 17 - 18 | 2396 | 83 |
BC151045.1 | - | 17 - 18 | 2392 | 83 | |
BC151053.1 | - | 17 - 18 | 2392 | 83 | |
EF570784.1 | - | 17 - 18 | 2305 | 83 |