Assay Design



Gene Transcripts

Gene Symbol
Kif19a
Entrez Gene ID
286942
Gene Name
kinesin family member 19A
Gene Aliases
Kif19
Chromosome Location
Chr.11: 114764501 - 114791118 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102615.1 NP_001096085.1 19 - 20 2993 86
GenBank mRNA AK155544.1 - 19 - 20 2959 86
BC151045.1 - 19 - 20 2955 86
BC151053.1 - 19 - 20 2955 86
EF570784.1 - 19 - 20 2868 86

View Full Product Details