Assay Design



Gene Transcripts

Gene Symbol
Slc17a8
Entrez Gene ID
216227
Gene Name
solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 8
Gene Aliases
BC042593, Vglut3
Chromosome Location
Chr.10: 89574020 - 89621249 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182959.3 NP_892004.1 1 - 2 451 80
GenBank mRNA AF510321.1 - 1 - 2 122 80
AK082743.1 - 1 - 2 197 80
BC042593.1 - 1 - 2 466 80

View Full Product Details