Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009408.2 | NP_033434.2 | 3 - 4 | 492 | 167 |
GenBank mRNA | AK145538.1 | - | 3 - 4 | 502 | 167 |
AK160590.1 | - | 3 - 4 | 346 | 167 | |
AK164061.1 | - | 3 - 4 | 411 | 167 | |
BC002095.1 | - | 3 - 4 | 373 | 167 | |
BC092349.1 | - | 3 - 4 | 322 | 167 | |
D10061.1 | - | 3 - 4 | 159 | 167 | |
L20632.1 | - | 3 - 4 | 159 | 167 |