Assay Design



Gene Transcripts

Gene Symbol
Eif5a
Entrez Gene ID
276770
Gene Name
eukaryotic translation initiation factor 5A
Gene Aliases
AA410058, D19Wsu54e, Eif4d, Eif5a1, eIF-4D, eIF-5A, eIF-5A-1, eIF-5A1
Chromosome Location
Chr.11: 69916712 - 69921958 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166589.1 NP_001160061.1 1 - 2 227 84
GenBank mRNA AY129326.1 - 1 - 2 227 84

View Full Product Details