Assay Design



Gene Transcripts

Gene Symbol
Trank1
Entrez Gene ID
320429
Gene Name
tetratricopeptide repeat and ankyrin repeat containing 1
Gene Aliases
A230061D21Rik, C030048J01Rik, Gm187, Lba1
Chromosome Location
Chr.9: 111311590 - 111395775 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164659.1 NP_001158131.1 19 - 20 5849 84
GenBank mRNA AK129119.1 - 7 - 8 1914 84
BC086653.1 - 7 - 8 875 84

View Full Product Details