Assay Design



Gene Transcripts

Gene Symbol
Trank1
Entrez Gene ID
320429
Gene Name
tetratricopeptide repeat and ankyrin repeat containing 1
Gene Aliases
A230061D21Rik, C030048J01Rik, Gm187, Lba1
Chromosome Location
Chr.9: 111311590 - 111395775 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164659.1 NP_001158131.1 6 - 7 1137 68
GenBank mRNA AK048832.1 - 6 - 7 941 68
AK079552.1 - 6 - 7 1138 68

View Full Product Details