Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019966.2 | NP_064350.2 | 3 - 4 | 852 | 78 |
GenBank mRNA | AK051564.1 | - | 3 - 4 | 803 | 78 |
AK082479.1 | - | 3 - 4 | 827 | 78 | |
AK171095.1 | - | 2 - 3 | 1686 | 78 | |
BC004764.1 | - | 3 - 4 | 852 | 78 |