Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019966.2 | NP_064350.2 | 4 - 5 | 1003 | 81 |
GenBank mRNA | AK051564.1 | - | 4 - 5 | 954 | 81 |
AK082479.1 | - | 4 - 5 | 978 | 81 | |
AK171095.1 | - | 3 - 4 | 1837 | 81 | |
BC004764.1 | - | 4 - 5 | 1003 | 81 | |
BC092290.1 | - | 1 - 2 | 174 | 81 |