Assay Design



Gene Transcripts

Gene Symbol
Etv4
Entrez Gene ID
18612
Gene Name
ets variant 4
Gene Aliases
AW414408, Pea-3, Pea3
Chromosome Location
Chr.11: 101769742 - 101785310 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008815.2 NP_032841.2 6 - 7 640 65
GenBank mRNA AK144538.1 - 6 - 7 600 65
AK144590.1 - 6 - 7 593 65
AK144683.1 - 4 - 5 354 65

View Full Product Details