Assay Design



Gene Transcripts

Gene Symbol
Etv4
Entrez Gene ID
18612
Gene Name
ets variant 4
Gene Aliases
AW414408, Pea-3, Pea3
Chromosome Location
Chr.11: 101769742 - 101785310 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008815.2 NP_032841.2 9 - 10 1141 83
GenBank mRNA AK144590.1 - 9 - 10 1094 83
AK144683.1 - 7 - 8 857 83
AK166136.1 - 9 - 10 1077 83
BC068248.1 - 1 - 2 1758 83
DQ832277.1 - 8 - 9 846 83

View Full Product Details