Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252193.1 | NP_001239122.1 | 2 - 3 | 481 | 76 |
NM_025275.4 | NP_079551.3 | 3 - 4 | 300 | 76 | |
GenBank mRNA | AK004062.1 | - | 3 - 4 | 260 | 76 |
AK075748.1 | - | 3 - 4 | 240 | 76 | |
AK143908.1 | - | 3 - 4 | 265 | 76 | |
BB872864.1 | - | 3 - 4 | 301 | 76 | |
BC025087.1 | - | 2 - 3 | 475 | 76 | |
CA575133.1 | - | 2 - 3 | 481 | 76 |