Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019650.3 | NP_062624.2 | 2 - 3 | 140 | 112 |
GenBank mRNA | AF007550.1 | - | 2 - 3 | 88 | 112 |
AK002556.1 | - | 2 - 3 | 130 | 112 | |
AK005697.1 | - | 2 - 3 | 118 | 112 | |
AK013052.1 | - | 2 - 3 | 134 | 112 | |
AK150004.1 | - | 2 - 3 | 138 | 112 | |
BC008525.1 | - | 2 - 3 | 136 | 112 | |
BC051253.1 | - | 2 - 3 | 106 | 112 |