Assay Design



Gene Transcripts

Gene Symbol
Exoc7
Entrez Gene ID
53413
Gene Name
exocyst complex component 7
Gene Aliases
Exo70, sec70
Chromosome Location
Chr.11: 116287998 - 116306749 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162872.1 NP_001156344.1 15 - 16 1749 85
NM_016857.2 NP_058553.2 17 - 18 1881 85
GenBank mRNA AF014461.1 - 17 - 18 1816 85
AK087601.1 - 15 - 16 1737 85
AK140450.1 - 17 - 18 1855 85
AK167769.1 - 15 - 16 1732 85
AK169928.1 - 15 - 16 1736 85
AK173094.1 - 15 - 16 6368 85
BC053710.1 - 15 - 16 1736 85

View Full Product Details