Assay Design



Gene Transcripts

Gene Symbol
Sgsm2
Entrez Gene ID
97761
Gene Name
small G protein signaling modulator 2
Gene Aliases
D630003G22Rik, R74628, Rutbc1, mKIAA0397
Chromosome Location
Chr.11: 74849262 - 74897093 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_197943.2 NP_922934.2 3 - 4 498 80
GenBank mRNA AK122273.1 - 3 - 4 420 80
BC058414.1 - 3 - 4 489 80
BC060163.1 - 3 - 4 489 80

View Full Product Details