Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004721.1 | NP_001004721.1 | 11 - 12 | 1219 | 75 |
GenBank mRNA | AK143517.1 | - | 11 - 12 | 1219 | 75 |
AK171988.1 | - | 11 - 12 | 1198 | 75 | |
AW538039.2 | - | 1 - 2 | 75 | ||
BC028278.1 | - | 3 - 4 | 350 | 75 | |
BC120680.1 | - | 11 - 12 | 1208 | 75 | |
BC120682.1 | - | 11 - 12 | 1208 | 75 | |
BE646884.1 | - | 1 - 2 | 75 |