Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004721.1 | NP_001004721.1 | 8 - 9 | 815 | 110 |
GenBank mRNA | AK143517.1 | - | 8 - 9 | 815 | 110 |
AK162913.1 | - | - | 790 | 110 | |
AK171988.1 | - | 8 - 9 | 794 | 110 | |
AW046539.1 | - | 3 - 4 | 296 | 110 | |
BC013076.1 | - | 1 - 2 | 106 | 110 | |
BC120680.1 | - | 8 - 9 | 804 | 110 | |
BC120682.1 | - | 8 - 9 | 804 | 110 |