Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291108.1 | NP_001278037.1 | 3 - 4 | 571 | 101 |
NM_025947.3 | NP_080223.2 | 3 - 4 | 327 | 101 | |
NR_111890.1 | 3 - 4 | 575 | 101 | ||
NR_111891.1 | 3 - 4 | 323 | 101 | ||
GenBank mRNA | AK002359.1 | - | 3 - 4 | 268 | 101 |
AK003435.1 | - | 2 - 3 | 257 | 101 | |
AK008214.1 | - | 3 - 4 | 288 | 101 | |
AK008595.1 | - | 4 - 5 | 289 | 101 | |
AK166685.1 | - | 3 - 4 | 303 | 101 | |
BC099423.1 | - | 3 - 4 | 287 | 101 | |
BC125610.1 | - | 3 - 4 | 287 | 101 | |
BC125648.1 | - | 3 - 4 | 287 | 101 | |
CO041603.1 | - | 1 - 2 | 101 |