Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145823.2 | NP_665822.1 | 8 - 9 | 1457 | 71 |
GenBank mRNA | AB077281.1 | - | 8 - 9 | 1438 | 71 |
AK171918.1 | - | 8 - 9 | 1454 | 71 | |
BC082333.1 | - | 8 - 9 | 1289 | 71 | |
BC108351.1 | - | 8 - 9 | 1317 | 71 |