Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010687.1 | NP_034817.1 | 14 - 15 | 2246 | 60 |
GenBank mRNA | AJ006278.1 | - | 14 - 15 | 2246 | 60 |
AK122328.1 | - | 15 - 16 | 3323 | 60 | |
AK133940.1 | - | 14 - 15 | 2731 | 60 | |
BC061506.1 | - | 13 - 14 | 2003 | 60 | |
BC079596.1 | - | 14 - 15 | 2527 | 60 | |
BC100399.1 | - | 14 - 15 | 2309 | 60 | |
BC125487.1 | - | 14 - 15 | 2212 | 60 | |
BC125489.1 | - | 14 - 15 | 2212 | 60 | |
HM141820.1 | - | 12 - 13 | 1881 | 60 |