Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010687.1 | NP_034817.1 | 7 - 8 | 1058 | 72 |
GenBank mRNA | AJ006278.1 | - | 7 - 8 | 1058 | 72 |
AK122328.1 | - | 8 - 9 | 2135 | 72 | |
AK133940.1 | - | 7 - 8 | 1542 | 72 | |
BC061506.1 | - | 6 - 7 | 815 | 72 | |
BC079596.1 | - | 7 - 8 | 1341 | 72 | |
BC100399.1 | - | 7 - 8 | 1121 | 72 | |
BC125487.1 | - | 7 - 8 | 1024 | 72 | |
BC125489.1 | - | 7 - 8 | 1024 | 72 | |
HM141820.1 | - | 5 - 6 | 693 | 72 |