Assay Design



Gene Transcripts

Gene Symbol
Lyl1
Entrez Gene ID
17095
Gene Name
lymphoblastomic leukemia 1
Gene Aliases
Lyl-1, bHLHa18
Chromosome Location
Chr.8: 84701422 - 84704716 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008535.2 NP_032561.2 1 - 2 535 70
GenBank mRNA AK076114.1 - 1 - 2 189 70
BC005736.1 - 1 - 2 155 70
X57687.1 - 1 - 2 445 70

View Full Product Details