Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001048145.1 | NP_001041610.1 | 1 - 2 | 91 | 69 |
NM_029139.1 | NP_083415.1 | 1 - 2 | 91 | 69 | |
GenBank mRNA | AK015606.1 | - | 1 - 2 | 87 | 69 |
AY899935.1 | - | 1 - 2 | 91 | 69 | |
AY899936.1 | - | 1 - 2 | 91 | 69 | |
BB616021.1 | - | 1 - 2 | 71 | 69 | |
BC141326.1 | - | 1 - 2 | 91 | 69 | |
BC141347.1 | - | 1 - 2 | 91 | 69 |