Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013384.2 | NP_001013402.2 | 7 - 8 | 879 | 98 |
GenBank mRNA | AK077813.1 | - | 2 - 3 | 1028 | 98 |
BC063764.1 | - | 7 - 8 | 879 | 98 | |
DQ303463.1 | - | 7 - 8 | 864 | 98 |