Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008261.2 | NP_032287.2 | 9 - 10 | 1404 | 74 |
GenBank mRNA | AK143948.1 | - | 9 - 10 | 1429 | 74 |
BC039220.1 | - | 9 - 10 | 1404 | 74 | |
D29015.1 | - | 9 - 10 | 1331 | 74 | |
FJ608820.1 | - | 10 - 11 | 1358 | 74 |