Assay Design
Gene Transcripts
- Gene Symbol
- Gnas
- Entrez Gene ID
-
14683
- Gene Name
- GNAS (guanine nucleotide binding protein, alpha stimulating) complex locus
- Gene Aliases
-
5530400H20Rik, A930027G11Rik, C130027O20Rik, GPSA, GSP, Galphas, Gnas1, Gnasxl, Gsa, Nesp, Nespl, Oed-Sml, Oedsml, P1, P2, P3, PHP1A, PHP1B, POH
- Chromosome Location
-
Chr.2: 174283444 - 174346744 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK039035.1 |
- |
2 - 3 |
1058 |
101 |
View Full Product Details