Assay Design



Gene Transcripts

Gene Symbol
Pfdn4
Entrez Gene ID
109054
Gene Name
prefoldin 4
Gene Aliases
0610025M10Rik, 1810063B06Rik, C-1, C1, Pfd4
Chromosome Location
Chr.2: 170496428 - 170519070 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013369.2 NP_001013387.1 1 - 2 170 107
GenBank mRNA BC065407.1 - 1 - 2 170 107
BC147673.1 - 1 - 2 71 107
BC147674.1 - 1 - 2 71 107

View Full Product Details