Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013369.2 | NP_001013387.1 | 3 - 4 | 417 | 82 |
NM_001110152.2 | NP_001103622.1 | 3 - 4 | 485 | 82 | |
NM_001199902.1 | NP_001186831.1 | 3 - 4 | 473 | 82 | |
GenBank mRNA | AK007940.1 | - | 3 - 4 | 278 | 82 |
AK136249.1 | - | 3 - 4 | 486 | 82 | |
BC048510.1 | - | 3 - 4 | 257 | 82 | |
BC065407.1 | - | 3 - 4 | 417 | 82 | |
BC147673.1 | - | 3 - 4 | 318 | 82 | |
BC147674.1 | - | 3 - 4 | 318 | 82 | |
BU503885.1 | - | 2 - 3 | 256 | 82 | |
CB523981.1 | - | 1 - 2 | 93 | 82 |