Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025499.2 | NP_079775.2 | 1 - 1 | 1148 | 76 |
GenBank mRNA | AK006438.1 | - | 1 - 1 | 1117 | 76 |
AK132745.1 | - | 1 - 1 | 1113 | 76 | |
AK161275.1 | - | 1 - 1 | 1148 | 76 | |
AK161307.1 | - | 1 - 1 | 1148 | 76 | |
BC099545.1 | - | 1 - 1 | 1115 | 76 |