Assay Design



Gene Transcripts

Gene Symbol
Arhgap40
Entrez Gene ID
545481
Gene Name
Rho GTPase activating protein 40
Gene Aliases
C20orf95, Gm14203, OTTMUSG00000015933
Chromosome Location
Chr.2: 158512796 - 158550628 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145015.1 NP_001138487.1 14 - 15 1934 84

View Full Product Details