Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178856.1 | NP_849187.1 | 3 - 4 | 359 | 71 |
GenBank mRNA | AK014776.1 | - | 3 - 4 | 373 | 71 |
AK028375.1 | - | 3 - 4 | 311 | 71 | |
AK030758.1 | - | - | 376 | 71 | |
BC082565.1 | - | 3 - 4 | 359 | 71 |