Assay Design



Gene Transcripts

Gene Symbol
Gins2
Entrez Gene ID
272551
Gene Name
GINS complex subunit 2 (Psf2 homolog)
Gene Aliases
2210013I18Rik, 4833427B12Rik, AI323585, Pfs2
Chromosome Location
Chr.8: 120581266 - 120589075 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178856.1 NP_849187.1 4 - 5 499 60
GenBank mRNA AK014776.1 - 4 - 5 513 60
AK030758.1 - - 516 60
BC082565.1 - 4 - 5 499 60

View Full Product Details