Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013519.2 | NP_038547.2 | 1 - 1 | 2649 | 109 |
GenBank mRNA | AI852431.1 | - | 1 - 1 | 109 | |
AK076319.1 | - | 1 - 1 | 2650 | 109 | |
AK146887.1 | - | 1 - 1 | 2651 | 109 | |
X74040.1 | - | 1 - 1 | 2624 | 109 | |
X92499.1 | - | 1 - 1 | 2231 | 109 |