Assay Design



Gene Transcripts

Gene Symbol
Pth1r
Entrez Gene ID
19228
Gene Name
parathyroid hormone 1 receptor
Gene Aliases
PPR, Pthr, Pthr1
Chromosome Location
Chr.9: 110722085 - 110747145 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011199.2 NP_035329.2 1 - 2 177 61
GenBank mRNA BC013446.1 - 1 - 2 154 61
BY005592.1 - 1 - 2 122 61

View Full Product Details