Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007462.1 | NP_001007463.1 | 5 - 6 | 695 | 68 |
GenBank mRNA | BC141316.1 | - | 5 - 6 | 694 | 68 |
BC141317.1 | - | 5 - 6 | 694 | 68 | |
BC145634.1 | - | 5 - 6 | 739 | 68 |