Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021513.2 | NP_067488.1 | 1 - 1 | 1703 | 76 |
GenBank mRNA | AB041579.1 | - | 1 - 1 | 1664 | 76 |
AK165981.1 | - | 1 - 1 | 1664 | 76 | |
AK166109.1 | - | 1 - 1 | 1700 | 76 | |
AK170457.1 | - | 1 - 1 | 1675 | 76 | |
AK210001.1 | - | 1 - 1 | 102 | 76 | |
BC003949.1 | - | 1 - 1 | 1256 | 76 |