Assay Design



Gene Transcripts

Gene Symbol
Bean1
Entrez Gene ID
65115
Gene Name
brain expressed, associated with Nedd4, 1
Gene Aliases
Bean
Chromosome Location
Chr.8: 104170376 - 104219098 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001141925.1 NP_001135397.1 1 - 2 494 97
GenBank mRNA BC030426.1 - 1 - 2 572 97

View Full Product Details