Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013380.2 | NP_001013398.2 | 10 - 11 | 1390 | 69 |
GenBank mRNA | AK140743.1 | - | 10 - 11 | 1158 | 69 |
AK147899.1 | - | 10 - 11 | 1157 | 69 | |
AK152157.1 | - | 10 - 11 | 1155 | 69 | |
AK161434.1 | - | 10 - 11 | 1252 | 69 | |
AK161494.1 | - | 10 - 11 | 1252 | 69 | |
AK207934.1 | - | 2 - 3 | 78 | 69 | |
BC058645.1 | - | 10 - 11 | 1348 | 69 |