Assay Design



Gene Transcripts

Gene Symbol
Ces2h
Entrez Gene ID
436059
Gene Name
carboxylesterase 2H
Gene Aliases
Ces2, EG436059, Gm5744
Chromosome Location
Chr.8: 105000853 - 105020410 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272045.1 NP_001258974.1 5 - 6 814 83

View Full Product Details