Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008290.2 | NP_032316.2 | 4 - 5 | 857 | 79 |
GenBank mRNA | AK004950.1 | - | 4 - 5 | 859 | 79 |
BC012682.1 | - | 4 - 5 | 846 | 79 | |
X95685.1 | - | 4 - 5 | 818 | 79 | |
Y09517.1 | - | 4 - 5 | 901 | 79 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | AI194967.1 | - | Chr. - - [-] on Build | 79 |