Assay Design



Gene Transcripts

Gene Symbol
Aar2
Entrez Gene ID
68295
Gene Name
AAR2 splicing factor homolog (S. cerevisiae)
Gene Aliases
0610011L14Rik
Chromosome Location
Chr.2: 156547558 - 156568972 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164818.1 NP_001158290.1 4 - 5 1475 86
NM_026661.4 NP_080937.3 3 - 4 1332 86
GenBank mRNA AK018741.1 - 3 - 4 1313 86
AK028890.1 - 3 - 4 1310 86
AK049107.1 - 2 - 3 733 86
AK166203.1 - 3 - 4 1313 86
AK169526.1 - 4 - 5 1467 86
AK210218.1 - 2 - 3 313 86
BC019988.1 - 3 - 4 1256 86

View Full Product Details