Assay Design



Gene Transcripts

Gene Symbol
Cc2d1a
Entrez Gene ID
212139
Gene Name
coiled-coil and C2 domain containing 1A
Gene Aliases
BC016188, Freud-1, Tape
Chromosome Location
Chr.8: 84132828 - 84147931 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145970.1 NP_666082.2 4 - 5 427 66
GenBank mRNA BC016188.1 - 4 - 5 427 66
BC027028.1 - 3 - 4 242 66

View Full Product Details