Assay Design



Gene Transcripts

Gene Symbol
Vat1l
Entrez Gene ID
270097
Gene Name
vesicle amine transport protein 1 homolog-like (T. californica)
Gene Aliases
9430073I07, AI427515, mKIAA1576
Chromosome Location
Chr.8: 114205582 - 114374074 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173016.3 NP_766604.2 4 - 5 794 82
GenBank mRNA AK122527.1 - 4 - 5 794 82
BC056927.1 - 4 - 5 804 82

View Full Product Details