Assay Design



Gene Transcripts

Gene Symbol
Vat1l
Entrez Gene ID
270097
Gene Name
vesicle amine transport protein 1 homolog-like (T. californica)
Gene Aliases
9430073I07, AI427515, mKIAA1576
Chromosome Location
Chr.8: 114205582 - 114374074 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173016.3 NP_766604.2 6 - 7 952 159
GenBank mRNA AK122527.1 - 6 - 7 952 159
AK189227.1 - 3 - 4 178 159
BC056927.1 - 6 - 7 962 159

View Full Product Details