Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146032.3 | NP_666144.3 | 11 - 12 | 1405 | 63 |
GenBank mRNA | AK033010.1 | - | 11 - 12 | 1324 | 63 |
AK076003.1 | - | 11 - 12 | 1302 | 63 | |
BC033573.1 | - | 7 - 8 | 705 | 63 | |
BC070422.1 | - | 11 - 12 | 1405 | 63 |