Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282108.1 | NP_001269037.1 | 5 - 6 | 686 | 92 |
NM_029736.2 | NP_084012.1 | 5 - 6 | 686 | 92 | |
NR_104094.1 | 4 - 5 | 603 | 92 | ||
GenBank mRNA | AK010834.1 | - | 5 - 6 | 92 | |
AK040517.1 | - | 5 - 6 | 565 | 92 | |
AK079901.1 | - | 5 - 6 | 663 | 92 | |
AK152752.1 | - | 5 - 6 | 647 | 92 | |
AK152849.1 | - | 5 - 6 | 647 | 92 | |
AK160402.1 | - | 5 - 6 | 670 | 92 | |
AY825926.1 | - | 5 - 6 | 439 | 92 | |
AY825927.1 | - | 4 - 5 | 356 | 92 | |
BC116666.1 | - | 5 - 6 | 552 | 92 | |
BC116717.1 | - | 5 - 6 | 552 | 92 |