Assay Design



Gene Transcripts

Gene Symbol
Ttc13
Entrez Gene ID
234875
Gene Name
tetratricopeptide repeat domain 13
Gene Aliases
BC017545
Chromosome Location
Chr.8: 124671327 - 124722013 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145607.3 NP_663582.2 16 - 17 1890 109
GenBank mRNA AK144842.1 - 16 - 17 1891 109
AK148102.1 - 18 - 19 2120 109
AK160894.1 - 18 - 19 2127 109
BC017545.1 - 10 - 11 1101 109
BC072577.1 - 16 - 17 1880 109
BC079851.1 - 17 - 18 1945 109

View Full Product Details